missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluticasone Furoate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Fluticasone Furoate
Specifications
Specifications
| Chemical Name or Material | Fluticasone Furoate |
| CAS | 397864-44-7 |
| Analyte or Component Names | Fluticasone Furoate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C27 H29 F3 O6 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Androsta-1,4-diene-17-carbothioic acid, 6,9-difluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxo-, S-(fluoromethyl) ester, (6α,11β,16α,17α)-, 6α,9α-Difluoro-17α-[(2-furanylcarbonyl)oxy]-11β-hydroxy-16α-methyl-3-oxoandrosta-1,4-diene-17β-carbothioic acid S-fluoromethyl ester, Allermist, Avamys, Fluticasone furoate, Furamist, GSK 685 698, GW 685698X, Veramyst, GW 685698X, CAF (GW694301X), GW 872664X, TAF (GW675713X), 6-des-fluoro 698 (GW836850X), Fluticasone Furoate (GW685698X), GSK465433A and GSK1322631A, 5-chloro-furoyl 698 (GSK180878A), TA-TAF dimer (GW871519X), GSK1042706A, GSK1103877A, GSK1322631A, Thioacid free acid (GR46580X), Thioacid furoate (GW675713X), Thioacid difuroate (GW725996X) |
| InChI Formula | InChI=1S/C27H29F3O6S/c1-14-9-16-17-11-19(29)18-10-15(31)6-7-24(18,2)26(17,30)21(32)12-25(16,3)27(14,23(34)37-13-28)36-22(33)20-5-4-8-35-20/h4-8,10,14,16-17,19,21,32H,9,11-13H2,1-3H3/t14-,16+,17+,19+,21+,24+,25+,26+,27+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)SCF |
| IUPAC Name | [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction