missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLVCR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 241.00 - € 326.00
Specifica
| Antigen | FLVCR |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
18647732
|
Novus Biologicals
NBP2-97925-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661752
|
Novus Biologicals
NBP2-97925-50ul |
50 μg |
€ 241.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
FLVCR Polyclonal antibody specifically detects FLVCR in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| FLVCR | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ataxia, posterior column 1, with retinitis pigmentosa, AXPC1, feline leukemia virus subgroup C cellular receptor 1, Feline leukemia virus subgroup C receptor, feline leukemia virus subgroup C receptor-related protein 1, FLVCRFLJ33420, hFLVCR, MFSD7B, PCA, PCARP | |
| Produced in rabbits immunized with E. coli-derived Human FLVCR fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 28982 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title