Learn More
FLYWCH2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24228-1-AP-150UL
 View more versions of this product
Product Code. 18068715
  • € 410.00 / 150µL
Description
The FLYWCH2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FLYWCH2. This antibody recognizes human antigen. The FLYWCH2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
FLYWCH2
700 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Immunohistochemistry, ELISA
FLYWCH family member 2, FLYWCH2
150 μL
114984
Store at -20°C. Stable for one year after shipment.