Learn More
FMC63 scFv Mouse, Unconjugated, Clone: Y45, AcroBiosystems
Description
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.
Specifications
Specifications
| Antigen | FMC63 scFv |
| Classification | Monoclonal |
| Clone | Y45 |
| Conjugate | Unconjugated |
| Host Species | Human (HEK293) |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | -20°C |
| Isotype | IgG1 |