missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMN1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00342184-B01P
This item is not returnable.
View return policy
Description
FMN1 Polyclonal antibody specifically detects FMN1 in Human samples. It is validated for Western Blot
Specifications
| FMN1 | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAI03693.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 342184 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| DKFZp686C2281, DKFZp686G2387, FLJ45135, FMNLimb deformity protein homolog, formin (limb deformity), formin 1, formin-1, LDDKFZP686C2281, MGC125288, MGC125289 | |
| FMN1 (AAI03693.1, 1 a.a. - 503 a.a.) full-length human protein. MENVDNSLDGSDVSEPAKPEAGLEVAQSILSKFSMKSLFGFTSKLESVNPEEEDAVLKAFHSLDVNPTSQQDDSSNGLDPQEAGSRVSPDLGNDEKIASVETESEGSQRKEAGTSLLAQELLPLSTLKGTKDDVICVRGTLVHTTSDSDSDDGGQEPEEGSSTNGPKSPSVVLSEPSQESKENPGGFRENTVTGEMNGAELCAEDPQRIPPEMSSKLEAGNGGLQTERRPSQDQVGEEGSQDLPAVTNQNSSVGITESASSKKEVSGEKSFQLPAFFSGLRVLKKGATAEGGETITEIKPKDGDLALLKLTQPVQKSLVQAGLQTVKSEKKATDPKATPTLLEQLSLLLNIDMPKTEPKGADPESPRREEMGCNADQESQSGPGVPQTQGGEVKPKSPETALEAFKALFIRPPRKGTTADTSELEALKRKMRHEKESLRAVFERSNSKPADGPSDSKSPDHSLTEQDDRTPGRLQAVWPPPKTKDTEEKVGLKYTEAGNVKGS | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction