missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FMN1 Monoclonal Antibody (4F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524537
This item is not returnable.
View return policy
Description
Peptide Sequence: MENVDNSLDG SDVSEPAKPE AGLEVAQSIL SKFSMKSLFG FTSKLESVNP EEEDAVLKAF HSLDVNPTSQ QDDSSNGLDP QEAGSRVSPD LGNDEKIASV ETESEGSQR.
Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
Specifications
| FMN1 | |
| Monoclonal | |
| Unconjugated | |
| Fmn1 | |
| FMN; Fmn1; formin (limb deformity); formin 1; formin-1; Ld; limb deformity protein; Limb deformity protein homolog; RGD1306349; RGD1310100 | |
| Mouse | |
| Protein A | |
| RUO | |
| 342184 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 4F4 | |
| PBS with no preservative; pH 7.4 | |
| Q68DA7 | |
| Fmn1 | |
| FMN1 (XP_375185.1, 1 a.a. ~ 109 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction