missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FMN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585687
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (87%). Positive Control: K562, THP-1, human brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FMN2, a novel formin homology protein of the cappuccino subfamily, is highly conserved between evolutionarily diverse vertebrates. FMN2 bears a high degree of similarity to formin-1 which is the founding member of formin homology proteins. FMN2 plays a crucial role in maintenance of the meiotic spindle. The protein is widely expressed in human fetal brain, adult whole brain, hypothalamus, retina, pancreatic islet and germinal-center B cells. Pathological role of the protein is identified in various tumors like parathyloid tumor, glioblastoma, retinoblastoma and chondrosarcoma. FMN2 mutations are often a cause of unexplained infertility in women. The gene encoding for human FMN2 is localized in the chromosomal region 1q43.
Specifications
| FMN2 | |
| Polyclonal | |
| Unconjugated | |
| FMN2 | |
| AU024104; FMN2; formin 2; formin-2; RP11-90L13.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56776 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.47 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9NZ56 | |
| FMN2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human FMN2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction