missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FMNL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30122
This item is not returnable.
View return policy
Description
FMNL1 Polyclonal antibody specifically detects FMNL1 in Human samples. It is validated for Immunoprecipitation
Specifications
| FMNL1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| C17orf1BMGC133052, C17orf1KW-13, CLL-associated antigen KW-13, FHOD4, FMNLMGC1894, formin-like, formin-like 1, formin-like protein 1, Leukocyte formin, MGC21878 | |
| Between 1025 and 1075 | |
| 100 μg | |
| Cancer, Immunology, Innate Immunity | |
| 752 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction