Learn More
FMNL2 Mouse, anti-Human, Clone: 3E8C12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68551-1-IG-20UL
 View more versions of this product
Product Code. 18014475
  • € 145.00 / 20µL
Description
The FMNL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FMNL2. This antibody recognizes human antigen. The FMNL2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
FMNL2
3E8C12
FHOD2, FLJ37546, FMNL2, formin like 2, Formin like protein 2, KIAA1902
20 μL
114793
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa