missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-86412-25ul
This item is not returnable.
View return policy
Description
FMO1 Polyclonal specifically detects FMO1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FMO1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| dimethylaniline monooxygenase [N-oxide-forming] 1, Dimethylaniline oxidase 1, EC 1.14.13.8, Fetal hepatic flavin-containing monooxygenase 1, flavin containing monooxygenase 1, Flavin-containing monooxygenase 1 (fetal liver), FMO 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2326 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FMO1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LPTPIVTWLMERKINNWLNHANYGLIPEDRTQLKEFVLNDELPGRIITGKVFIRPSIKEVKENSVIFNNTSKEEPIDIIVFATGYTFAFPFLDESVVKVEDG | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido