Learn More
Invitrogen™ FMO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109732
Description
Immunogen sequence: VFISTRHGTW VMSRISEDGY PWDSVFHTRF RSMLRNVLPR TAVKWMIEQQ MNRWFNHENY GLEPQNKYIM KEPVLNDDVP SRLLCGAIKV KSTVKELTET SAIFEDGTVE ENIDVIIFAT GYSFSFPFLE DSLVKVENNM VSLYKYIFPA HLDKSTLACI GLIQPLGSIF PTAELQARWV TRVFKGLCSL PSERTMMMDI IKRNEKRIDL FGESQSQTLQ TNYVDYLDEL ALEIGAKPDF CSLLFKDPKL AVRLYFGPCN S.
The flavin-containing monooxygenases are NADPH-dependent enzymes that catalyze the oxidation of many drugs and xenobiotics. In most mammals, there is a flavin-containing monooxygenase that catalyzes the N-oxidation of some primary alkylamines through an N-hydroxylamine intermediate. However, in humans, this enzyme is truncated and is probably rapidly degraded. The protein encoded by this gene represents the truncated form and apparently has no catalytic activity. A functional allele found in African Americans has been reported, but no sequence evidence has been deposited to support the finding. This gene is found in a cluster with the FMO1, FMO3, and FMO4 genes on chromosome 1.
Specifications
| FMO2 | |
| Polyclonal | |
| Unconjugated | |
| Fmo2 | |
| 2310008D08Rik; 2310042I22Rik; AW107733; Dimethylaniline monooxygenase [N-oxide-forming] 2; dimethylaniline oxidase 2; flavin containing dimethylaniline monoxygenase 2; flavin containing monooxygenase 2; flavin containing monooxygenase 2 (non-functional); FMO 1B1; FMO 2; FMO, pulmonary; FMO1B1; FMO2; Pulmonary flavin-containing monooxygenase 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2327 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.66 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99518 | |
| Fmo2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-470 of human FMO2 (NP_001451.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.