missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-(R)-3-amino-3-(4-hydroxyphenyl)propionic Acid, TRC

Product Code. 30402656
Change view
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402656 1 g 1g
30399332 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402656 Supplier Toronto Research Chemicals Supplier No. TRCF6575601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fmoc-(R)-3-amino-3-(4-hydroxyphenyl)propionic Acid
CAS 511272-36-9
Molecular Formula C24 H21 N O5
Quantity 1 g
Synonym (βR)-β-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-hydroxy-benzenepropanoic Acid
InChI Formula InChI=1S/C24H21NO5/c26-16-11-9-15(10-12-16)22(13-23(27)28)25-24(29)30-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22,26H,13-14H2,(H,25,29)(H,27,28)/t22-/m1/s1
SMILES OC(=O)C[C@@H](NC(=O)OCC1c2ccccc2c3ccccc13)c4ccc(O)cc4
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 403.427
Formula Weight 403.142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.