Learn More
Fmoc-(R)-3-amino-3-(4-hydroxyphenyl)propionic Acid, TRC
Product Code. 30391997
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391997 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30391997 Supplier Toronto Research Chemicals Supplier No. TRCF657560250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fmoc-(R)-3-amino-3-(4-hydroxyphenyl)propionic Acid
CAS 511272-36-9
Molecular Formula C24 H21 N O5
Quantity 250 mg
Synonym (βR)-β-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-hydroxy-benzenepropanoic Acid
InChI Formula InChI=1S/C24H21NO5/c26-16-11-9-15(10-12-16)22(13-23(27)28)25-24(29)30-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22,26H,13-14H2,(H,25,29)(H,27,28)/t22-/m1/s1
SMILES OC(=O)C[C@@H](NC(=O)OCC1c2ccccc2c3ccccc13)c4ccc(O)cc4
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 403.427
Formula Weight 403.142