Learn More
FMR1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1C6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504290
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FMR1 binds RNA and is associated with polysomes. The protein may be involved in mRNA trafficking from the nucleus to the cytoplasm. A trinucleotide repeat in the 5′ UTR is normally found at 6-53 copies, but an expansion to 55-230 repeats is the cause of fragile X syndrome. Expansion of the trinucleotide repeat may also cause one form of premature ovarian failure. Multiple alternatively spliced transcript variants that encode different protein isoforms and which are located in different cellular locations have been described for this gene.Specifications
| FMR1 | |
| Monoclonal | |
| Unconjugated | |
| FMR1 | |
| FMRP, FRAXA, POF, POF1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 14265, 2332, 24948, 481070 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P35922, Q06787, Q80WE1 | |
| Fmr1 | |
| Human recombit protein fragment corresponding to amino acids 36-279 of human FMR1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.