missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNBP1L Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76845IR
This item is not returnable.
View return policy
Description
FNBP1L Polyclonal antibody specifically detects FNBP1L in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FNBP1L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| C1orf39, chromosome 1 open reading frame 39, FLJ20275, formin binding protein 1-like, formin-binding protein 1-like, toca-1, TOCA1Transducer of Cdc42-dependent actin assembly protein 1, transducer of Cdc42-dependent actin assembly 1 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human TOCA-1. The immunogen is located within the last 50 amino acids of TOCA-1. Amino Acid Squence: GWTRARRQNGEEGYVPT | |
| 0.1 mL | |
| Neuroscience | |
| 54874 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction