missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FNBP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103846
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FNBP4.
FNBP4 encodes a protein containing two tryptophan-rich WW domains that binds the proline-rich formin homology 1 domains of formin family proteins, suggesting a role in the regulation of cytoskeletal dynamics during cell division and migration. It also binds intersectin family proteins suggesting a role in the maintenance of membrane curvature at sites of nascent vesicle formation. Naturally occurring mutations in this gene are associated with Waardenburg anophthalmia syndrome.
Specifications
| FNBP4 | |
| Polyclonal | |
| Unconjugated | |
| FNBP4 | |
| FBP30; Fnbp30; Fnbp4; formin binding protein 30; formin binding protein 4; formin-binding protein 30; Formin-binding protein 4; Kiaa1014; mKIAA1014 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23360, 55935 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ZQ03, Q8N3X1 | |
| FNBP4 | |
| A synthesized peptide derived from human FNBP4(Accession Q8N3X1), corresponding to amino acid residues Q983-T1017. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction