missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Apolipoprotein C1 ELISA Kit (Colorimetric)

Product Code. 18684430 Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Kit
missing translation for 'unitSize'
1 set
This item is not returnable. View return policy

Product Code. 18684430

missing translation for 'mfr': Novus Biologicals™ NBP260611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.031 to 2 μg/mL (example only, lot dependent)
Assay Sensitivity 0.01 μg/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Culture
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 341
Gene Symbol APOC1
Quantity 1 Kit
Research Discipline Cancer, Lipid and Metabolism
Storage Requirements Storage is content dependent.
Synonym APOC1, APOC1B, Apo-CIB, ApoC-IB, Apolipoprotein C1, apolipoprotein C-I
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.