missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNDC4 Antibody (OTI1B7), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72151AF488
This item is not returnable.
View return policy
Description
FNDC4 Monoclonal specifically detects FNDC4 in Human, Mouse samples. It is validated for Immunohistochemistry.
Specifications
| FNDC4 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FNDC4 | |
| Full length recombinant protein of human FNDC4 (NP_073734) produced in HEK293T cells. | |
| 0.1 mL | |
| 64838.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI1B7 | |
| Immunohistochemistry | |
| fibronectin type III domain containing 4, fibronectin type III domain-containing protein 4, Fibronectin type III repeat-containing protein 1, FRCP1FLJ22362 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction