missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNDC4 Antibody (OTI1B7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72151CL1
This item is not returnable.
View return policy
Description
FNDC4 Monoclonal antibody specifically detects FNDC4 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| FNDC4 | |
| Monoclonal | |
| CoraFluor 1 | |
| fibronectin type III domain containing 4, fibronectin type III domain-containing protein 4, Fibronectin type III repeat-containing protein 1, FRCP1FLJ22362 | |
| Full length human recombinant protein of human FNDC4(NP_073734) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1B7 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 64838 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction