missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FNDC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579279
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat heart tissue, mouse heart tissue.
Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.
Specifications
| FNDC5 | |
| Polyclonal | |
| Unconjugated | |
| FNDC5 | |
| 1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 252995, 260327, 384061 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q8K3V5, Q8K4Z2, Q8NAU1 | |
| FNDC5 | |
| E. coli-derived human FNDC5 recombinant protein (Position: D32-E143). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction