missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNIP1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18249DL594
This item is not returnable.
View return policy
Description
FNIP1 Polyclonal antibody specifically detects FNIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| FNIP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp781P0215, folliculin interacting protein 1, folliculin-interacting protein 1, KIAA1961DKFZp686E18167, MGC667 | |
| Synthetic peptide from the mid-protein of Human FNIP1 | |
| 0.1 mL | |
| Cell Biology | |
| 96459 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction