missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNIP2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81934AF350
This item is not returnable.
View return policy
Description
FNIP2 Polyclonal antibody specifically detects FNIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FNIP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 57600 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| FNIP1-like protein, FNIPL, folliculin interacting protein 2 | |
| FNIP2 antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human FNIP2. The immunogen is located within amino acids 860 - 910 of FNIP2. Amino Acid Squence: KKANFRTEGDIPRNES | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction