Learn More
Motic™ Widefield Eyepiece with Diopter Adjustment for AE Microscope
Focusable eyepiece increases magnification. Motic™ Widefield Eyepiece with Diopter Adjustment is for use with an inverted AE microscope.
Brand: Motic™ 1101001402061
Additional Details : Weight : 0.80000kg
Description
- For use with AE Series Microscopes
- Magnification/Field Number: 12.5X/18mm or 15X/16mm
- Includes Diopter Adjustment
Order Info
dimProductType
Specifications
Focusable Widefield Eyepiece | |
16 mm | |
15X | |
1 Ea. |