missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOG2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | FOG2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18393196
|
Novus Biologicals
NBP3-03827-20UL |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18372997
|
Novus Biologicals
NBP3-03827-100UL |
100 μL |
€ 470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOG2 Polyclonal specifically detects FOG2 in Human, Mouse samples. It is validated for Western Blot.Specifications
| FOG2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 23414 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| transcription factor GATA4, modulator of, zinc finger protein 89B, zinc finger protein, multitype 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 982-1151 of human FOG2 (NP_036214.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title