missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid Hydrate EP Impurity B (as Sulfate), Mikromol™

Product Code. 30457417
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457417 4 x 25 mg 100mg
30049329 100 mg 100mg
30049220 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457417 Supplier Mikromol Supplier No. MM0621.0200254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Folic Acid Hydrate EP Impurity B (as Sulfate), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5,6-Triaminopyrimidin-4(1H)-one Sulphate
CAS 39267-74-8
Analyte or Component Names 2,5,6-Triaminopyrimidin-4(1H)-one Sulphate
Shipping Condition Room Temperature
Molecular Formula C4 H7 N5 O . H2 O4 S
Certifications/Compliance ISO 9001
Synonym 2,5,6-Triaminopyrimidin-4(1H)-one Sulphate, Folic Acid Imp. B (EP) as Sulphate, Folic Acid Hydrate Impurity B as Sulfate, Folic Acid Impurity B as Sulfate
InChI Formula InChI=1S/C4H7N5O.H2O4S/c5-1-2(6)8-4(7)9-3(1)10;1-5(2,3)4/h5H2,(H5,6,7,8,9,10);(H2,1,2,3,4)
Solution Type Neat
SMILES NC1=NC(=O)C(=C(N)N1)N.OS(=O)(=O)O
IUPAC Name sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one
Molecular Weight (g/mol) 239.21
Quantity 4 x 25 mg
Formula Weight 239.0324 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.