Learn More
Novus Biologicals™ Folliculin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30658
Product Code. 30044379
  • € 571.00 / 100µg
Description
Folliculin Polyclonal antibody specifically detects Folliculin in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Folliculin
Polyclonal
Tris-citrate/phosphate buffer, pH 7 to 8
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
BHD skin lesion fibrofolliculoma protein, BHDFLCL, Birt-Hogg-Dube syndrome protein, DKFZp547A118, FLJ45004, FLJ99377, folliculin, MGC17998, MGC23445
between 1 and 50
100 μg
Cancer, Cell Cycle and Replication
201163
Store at 4°C. Do not freeze.