missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Folliculin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555971
This item is not returnable.
View return policy
Description
Immunogen sequence: VGCEDDQSLS KYEFVVTSGS PVAADRVGPT ILNKIEAALT NQNLSVDVVD QCLVCLKEEW MNKVKVLFKF TKVDSRPKED TQKLLSILGA SEEDNVKLLK FWMTGLSKTY KSHLMSTVR Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
Specifications
| Folliculin | |
| Polyclonal | |
| Unconjugated | |
| FLCN | |
| AU014660; B430214A04Rik; BHD; BHD skin lesion fibrofolliculoma protein; birt-Hogg-Dube syndrome protein; Birt-Hogg-Dube syndrome protein homolog; FLCL; Flcn; Folliculin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 201163 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFG4 | |
| FLCN | |
| Recombinant protein corresponding to Human Folliculin. Recombinant protein control fragment (Product #RP-93335). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction