missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Folliculin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100247
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FLCN.
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
Specifications
| Folliculin | |
| Polyclonal | |
| Unconjugated | |
| FLCN | |
| AU014660; B430214A04Rik; BHD; BHD skin lesion fibrofolliculoma protein; birt-Hogg-Dube syndrome protein; Birt-Hogg-Dube syndrome protein homolog; FLCL; Flcn; Folliculin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 201163, 216805, 303185 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q76JQ2, Q8NFG4, Q8QZS3 | |
| FLCN | |
| A synthesized peptide derived from human FLCN(Accession Q8NFG4), corresponding to amino acid residues S56-Y106. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction