missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR1 Antibody (mirvetuximab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28386CL1
This item is not returnable.
View return policy
Description
FOLR1 Monoclonal antibody specifically detects FOLR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| FOLR1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, FOLR, FR-alpha, KB cells FBP, Ovarian tumor-associated antigen MOv18 | |
| FOLR1/ FRA | |
| 0.1 mL | |
| Cancer | |
| 2348 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| mirvetuximab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction