missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR2 Antibody (046), DyLight 594 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89847DL594
This item is not returnable.
View return policy
Description
FOLR2 Monoclonal antibody specifically detects FOLR2 in Human samples. It is validated for ELISA
Specifications
| FOLR2 | |
| Monoclonal | |
| DyLight 594 | |
| BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FOLR2 (P14207-1; Met 1-His 228). | |
| 0.1 mL | |
| Cancer | |
| 2350 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 046 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction