missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR2 Antibody (OTI4G6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70763AF350
This item is not returnable.
View return policy
Description
FOLR2 Monoclonal specifically detects FOLR2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FOLR2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FOLR2 | |
| Recombinant protein fragment corresponding to amino acids 20-221 of human FOLR2 (NP_000794) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4G6 | |
| Western Blot, Immunohistochemistry | |
| BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
| Mouse | |
| 0.1 mL | |
| 2350 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction