Få mere at vide
FOLR2 Mouse anti-Human, Clone: OTI4G6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808026
Beskrivelse
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FOLR2 is a member of the folate receptor (FOLR) family, and these genes exist in a cluster on chromosome 11. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and they mediate delivery of 5-methyltetrahydrofolate to the interior of cells. This protein has a 68% and 79% sequence homology with the FOLR1 and FOLR3 proteins, respectively. Although this protein was originally thought to be specific to placenta, it can also exist in other tissues, and it may play a role in the transport of methotrexate in synovial macrophages in rheumatoid arthritis patients.Tekniske data
| FOLR2 | |
| Monoclonal | |
| Unconjugated | |
| FOLR2 | |
| BETA-HFR, FBP, FBP/PL-1, FR-BETA, FR-P3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2350 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P14207 | |
| FOLR2 | |
| Human recombit protein fragment corresponding to amino acids 20-221 of human FOLR2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.