missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR4 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99508C
This item is not returnable.
View return policy
Description
FOLR4 Polyclonal antibody specifically detects FOLR4 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| FOLR4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 390243 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| DyLight 650 | |
| folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FOLR4. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction