missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR4 Antibody (TH6), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-26657G
This item is not returnable.
View return policy
Description
FOLR4 Monoclonal specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| FOLR4 | |
| Monoclonal | |
| DyLight 488 | |
| folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta | |
| Rat | |
| Protein G purified | |
| RUO | |
| 390243 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Flow Cytometry | |
| TH6 | |
| Flow Cytometry 1ul/1 million cells | |
| FOLR4 | |
| CD3 mAb-stimulated CD4/25 T cells | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction