missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR4 Antibody (TH6), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-26657F
This item is not returnable.
View return policy
Description
FOLR4 Monoclonal specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| FOLR4 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| IZUMO1R | |
| CD3 mAb-stimulated CD4/25 T cells | |
| 0.1 mL | |
| 390243 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| TH6 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta | |
| Rat | |
| Protein G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction