Learn More
MilliporeSigma Foot Pedal for Millicell™ ERS 3.0
Product Code. 18860309 Shop All Merck Products
Change view
Click to view available options
Voltage:
0/1 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
18860309 0/1 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18860309 Supplier MilliporeSigma Supplier No. MERS03PED
  • ONLINE EXCLUSIVE PRICE
    € 208.85
    € 199.63 / Pack of 1
    Save € 9.22
    4% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Optional external foot pedal or switch for Millicell ERS 3.0 Voltohmmeter.
  • The Millicell™ ERS 3.0 Digital VoltOhmMeter reliably measures membrane potential and resistance of epithelial and endothelial cells in culture. This device qualitatively measures cell monolayer health and cell confluence. The foot switch/pedal accessory can be attached to the Millicell™ ERS 3.0 system to achieve hands-free operation.
TRUSTED_SUSTAINABILITY
Specifications
Type Foot Pedal
Voltage 0/1 V