Learn More
Hirschmann™ Plastic Base and Safety Ring
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
Each
Description
- For measuring cylinders
- Without base
- Available to fit a range of sizes
Specifications
Specifications
| Material | Plastic |
| Product Type | Foot With Protective Ring |
| For Use With (Application) | For measuring cylinder |
| Quantity | 250 mL |