missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH FORTUNA™ Borosilicate Glass Class A Volumetric Flasks

Created from borosilicate glass. Poulten Graf™ FORTUNA™ Borosilicate Glass Class A Volumetric Flasks offers excellent chemical resistance and are best for obtaining a precise measurement of chemical solutions.

Brand:  Poulten & Graf GmbH 1 51646F

Additional Details : Weight : 1.25000kg

 View more versions of this product

Product Code. 15982270

  • € 145.00 / Pack of 5
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • Volumetric flasks
  • Class A
  • Amber color
Specifications

Specifications

134°C
White Graduation
Borosilicate Glass
Luer-lock
A
NS 14/23
Polypropylene, Glass
50 mL
134°C
Yes
Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers