Learn More
Poulten & Graf GmbH FORTUNA™ Bulb Pipette with Suction Piston
Product Code. p-7116402
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
Tolerance:
0.008 mL
0.010 mL
0.015 mL
0.020 mL
0.03 mL
0.030 mL
0.05 mL
Graduation Color:
Amber
Blue
Unit Size:
Each
Product Code. 10588521
Brand: Poulten & Graf GmbH 1 2821902
Only null left
Add to basket
Add to basket
Eliminate the need for a pipetting aid. Poulten & Graf GmbH FORTUNA™ Bulb Pipette with Suction Piston ensures reliable liquid dispensing from the pipette.
  • Includes 1 mark
  • Clear calibration cylinders
  • No pipetting aid necessary
  • Without certificate
Specifications
Class A
Material Glass
Sterility Non-sterile
Graduation Color Blue
Quantity 1
Tolerance 0.008 mL
Volume (Metric) 1 mL
Type Bulb Pipette with suction piston
Disposable Reusable