Learn More
Poulten & Graf GmbH FORTUNA™ Burette Bang pattern
Product Code. 10771693
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10771693 10 mL Each
10331462 5 mL Each
10341462 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10771693 Supplier Poulten & Graf GmbH Supplier No. 1 03034G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve better accuracy and repeatability, especially in titrations. Poulten & Graf GmbH FORTUNA™ Burette Bang pattern delivers accurate and precise volumes.
  • Comformity approved
  • Blue graduation
  • With glass key in upper stopcock
  • With straight glass stopcock
Specifications
Capacity (Metric) 10 mL
Material Borosilicate Glass
Graduations 0.02 mL
For Use With (Application) Burette Bang Pattern
Scale Blue
Stopcock Straight
Stopcock Material Glass
Tolerance ±0.02 mL
Class Class AS
Certifications/Compliance EN ISO 385