Learn More
Poulten Graf™ Fortuna™ Class A Glass Blue Color Graduation Pipettes
Provides volumetric accuracy for your pipetting needs. Poulten Graf™ Fortuna™ Class A Glass Blue Color Graduation Pipettes are manufactured from clear glass with permanent blue graduations.
Brand: Poulten Graf™ 1 1423202
Additional Details : Weight : 0.06300kg
Description
- Resistant against acids and bases
- Accuracy better than Class B
- Pipette aid not necessary
Specifications
A | |
0.05 mL | |
Clear | |
Graduated Pipette |
Yes | |
1 Pack | |
Glass | |
5 mL |