Learn More
Poulten Graf™ FORTUNA™ Class B Low-Form Glass Measuring Cylinders
€ 9.10 - € 75.80
Specifications
Base Type Hexagonal
Calibration To Contain
Certifications/Compliance EN ISO 4788
Class B
Type Graduated Measuring Cylinder
View More Specs
Products 7
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
15942150
View Documents Promotion Details
Poulten Graf™
1 3204502
50 mL 2 mL
€ 18.85
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
15911371
View Documents Promotion Details
Poulten Graf™
1 3205902
500 mL 10 mL
€ 75.80
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
15972150
View Documents Promotion Details
Poulten Graf™
1 3205502
250 mL 5 mL
€ 39.45
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
10428421
View Documents Promotion Details
Poulten Graf™
1 3205102
100 mL 2 mL
€ 20.40
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 22-04-2024
to see stock.
Add to basket
10269761
View Documents Promotion Details
Poulten Graf™
1 3204502
50 mL 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10209671
View Documents Promotion Details
Poulten Graf™
1 3205502
250 mL 5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10791113
View Documents Promotion Details
Poulten Graf™
1 3205902
500 mL 10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications
Hexagonal
EN ISO 4788
Graduated Measuring Cylinder
Round
Blue Graduations
To Contain
B
Blue
Borosilicate Glass