Learn More
Poulten & Graf GmbH FORTUNA™ Mixing Cylinders with PP-Stopper
Poulten & Graf GmbH FORTUNA™ Mixing Cylinders with PP-Stopper measures the volume of a liquid using horizontal marked lines.
€ 15.30 - € 184.00
Specifications
Base Type | Hexagonal |
---|---|
Calibration | To Contain |
Shape | Round |
Type | Graduated Mixing Cylinder |
Material | Borosilicate Glass |
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
15982170
|
Poulten & Graf GmbH
1 4205102 |
100 mL | 1 mL |
€ 42.85
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15912180
|
Poulten & Graf GmbH
1 4205504 |
250 mL | 2 mL |
€ 69.55
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15932170
|
Poulten & Graf GmbH
1 4203704 |
10 mL | 0.2 mL |
€ 31.90
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15992170
|
Poulten & Graf GmbH
1 4205104 |
100 mL | 1 mL |
€ 42.85
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15972170
|
Poulten & Graf GmbH
1 4204504 |
50 mL | 1 mL |
€ 38.70
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15962170
|
Poulten & Graf GmbH
1 4204502 |
50 mL | 1 mL |
€ 38.70
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15952180
|
Poulten & Graf GmbH
1 4206104 |
1000 mL | 10 mL |
€ 184.00
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15952170
|
Poulten & Graf GmbH
1 4204304 |
25 mL | 0.5 mL |
€ 33.80
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
10417271
|
Poulten & Graf GmbH
1 4203704 |
10 mL | 0.2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10016191
|
Poulten & Graf GmbH
1 4204504 |
50 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10535882
|
Poulten & Graf GmbH
1 4205102 |
100 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10417081
|
Poulten & Graf GmbH
1 4204502 |
50 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10679362
|
Poulten & Graf GmbH
1 4205504 |
250 mL | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10729012
|
Poulten & Graf GmbH
1 4206104 |
1000 mL | 10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10006191
|
Poulten & Graf GmbH
1 4205104 |
100 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10670323
|
Poulten & Graf GmbH
1 4204304 |
25 mL | 0.5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Conformity approved
- Hexagonal base
- With PP-stopper
Specifications
Hexagonal | |
Round | |
Borosilicate Glass | |
Class A | |
Polypropylene Stopper | |
Polypropylene | |
Autoclavable |
To Contain | |
Graduated Mixing Cylinder | |
EN ISO 4788 | |
Non-Disposable | |
1 | |
Polypropylene |