Learn More
Poulten & Graf GmbH FORTUNA™ OPTIFIX™ Safety S Dispenser
Click to view available options
Volume (Metric):
20 to 100 mL
Unit Size:
Each
Description
- For dispensing strong fuming acids and very abrasice liquids
- No metal spring
- No contamination of the dosing liquid
Specifications
Specifications
| Accuracy | 0.6% |
| Product Type | Bottletop Dispenser |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | EN ISO 8655-5 |
| Description | OPTIFIX SAFETY S Dispenser |
| Material | PTFE |
| Volume (Metric) | 20 to 100 mL |
| Includes | 2 PTFE Adaptors GL 40 and GL 45 |
| Increments | 2 mL |
| Model | OPTIFIX SAFETY S Dispenser |
| Show More |