Learn More
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Luer-Lock Tip
Product Code. 10333951
Click to view available options
Volume (Metric):
20 mL
30 mL
Unit Size:
Each
Product Code. 10333951
Brand: Poulten & Graf GmbH 7 24041
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Luer-Lock Tip is used to measure and dispense fluids.
  • With Luer-lock tip
  • With steel piston
  • Interchangable
  • For technical use only
Specifications
Autoclavable Autoclavable
Material Neutral Glass, Neutral Glass (Barrel), Nickel-plated Brass (Tip), Steel (Plunger)
Color Clear
Description Glass and Metal Syringe with Luer-Lock tip
Latex Free Yes
Product Type Glass and Metal Syringe
Disposable Reusable
Graduations Amber
Model Glass and Metal Syringe
Quantity 1
Volume (Metric) 20 mL
Connection Type Luer Lock
Graduated Yes