missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH FORTUNA™ OPTIMAT™ 3 Dispensing Pump

Poulten & Graf GmbH FORTUNA™ OPTIMAT™ 3 Dispensing Pump attaches to an OPTIMAT™ dispenser.

Brand:  Poulten & Graf GmbH 101 12653

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10462501

  • € 1595.00 / Each
Estimated Shipment: 29-04-2024
to see stock.
Add to basket

Description

Description

  • For dispensing strong fuming acids and very abrasive liquids
  • Intake and discharge tube sets are not included
  • For dispensing station
Specifications

Specifications

0.6%
Autoclavable
0.2%
200 mL
1
Dispensing Pumps for Automatic Station
EN ISO 8655
Dispensing pump for OPTIMAT automatic dispensing station
OPTIMAT automatic dispensing station
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers