Learn More
Poulten & Graf GmbH FORTUNA™ OPTIMAT™ 3 Dispensing Pump
Product Code. p-8498023
Click to view available options
Volume (Metric):
2 mL
10 mL
30 mL
50 mL
200 mL
300 mL
Unit Size:
Each
Product Code. 10156570
Brand: Poulten & Graf GmbH 101 12648
Only null left
Add to basket
Add to basket
Poulten & Graf GmbH FORTUNA™ OPTIMAT™ 3 Dispensing Pump attaches to an OPTIMAT™ dispenser.
  • For dispensing strong fuming acids and very abrasive liquids
  • Intake and discharge tube sets are not included
  • For dispensing station
Specifications
Accuracy 0.6%
Product Type Dispensing Pumps for Automatic Station
Autoclavable Autoclavable
Certifications/Compliance EN ISO 8655
Coefficient of Variation 0.2%
Description Dispensing pump for OPTIMAT automatic dispensing station
Volume (Metric) 50 mL
For Use With (Equipment) OPTIMAT automatic dispensing station
Quantity 1