Learn More
Poulten & Graf GmbH FORTUNA™ Precision Gas Syringe
Product Code. 10333891
Click to view available options
Volume (Metric):
25 mL
50 mL
100 mL
Includes:
3-way Stopcock
Capillary Tube
Straight Stopcock
Unit Size:
Each
Product Code. 10333891
Brand: Poulten & Graf GmbH 2 01251
Poulten & Graf GmbH FORTUNA™ Precision Gas Syringe are used to measure gas volume.
  • Clear neutral glass
  • Calibrated cylinders
  • Acid and alkali resistant amber stain diffused graduation
  • Amber graduation
Specifications
Autoclavable Autoclavable
Material Glass (Tip), Neutral Glass, Neutral Glass (Barrel), Neutral Glass (Plunger)
Color Clear
Includes Straight Stopcock
Product Type FORTUNA Precision Gas Syringe with straight stopcock
Disposable Reusable
Graduations Amber
Quantity 1
Volume (Metric) 100 mL
Graduated Yes
Sterility Non-sterile