missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOSB Monoclonal Antibody (OTI1A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804452
This item is not returnable.
View return policy
Description
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| FOSB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P53539 | |
| FOSB | |
| Human recombit protein fragment corresponding to amino acids 1-255 of human FOSB produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1A7 | |
| Unconjugated | |
| FOSB | |
| AP-1, G0S3, GOS3, GOSB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2354 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu