missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOSB Monoclonal Antibody (OTI6E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804286
This item is not returnable.
View return policy
Description
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| FOSB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P53539 | |
| FOSB | |
| Human recombit protein fragment corresponding to amino acids 1-255 of human FOSB produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI6E4 | |
| Unconjugated | |
| FOSB | |
| AP-1, G0S3, GOS3, GOSB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2354 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction